Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ncx-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269165.1 | NP_001256094.1 | 5 - 6 | 813 | 76 | |
NM_001373058.1 | NP_001360010.1 | 6 - 7 | 768 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |