Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: air-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072718.10 | NP_505119.1 | 1 - 2 | 61 | 71 | |
GenBank mRNA | AF071206.1 | - | 1 - 2 | 78 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |