Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srsx-22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072752.3 | NP_505153.2 | 4 - 5 | 577 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |