Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mom-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072753.6 | NP_505154.1 | 1 - 2 | 219 | 102 | |
GenBank mRNA | AF013952.1 | - | 1 - 2 | 183 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |