Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mom-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072753.6 | NP_505154.1 | 2 - 3 | 344 | 88 | |
GenBank mRNA | AF013952.1 | - | 2 - 3 | 308 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |