Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpa-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072756.5 | NP_505157.1 | 7 - 8 | 744 | 85 | |
GenBank mRNA | AY008125.1 | - | 7 - 8 | 735 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |