Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcy-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028783.1 | NP_001023954.1 | 17 - 18 | 2619 | 96 | |
NM_001028784.1 | NP_001023955.1 | 17 - 18 | 2619 | 96 | ||
NM_001373061.1 | NP_001360539.1 | 17 - 18 | 2619 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |