Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: otpl-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072795.4 | NP_505196.2 | 7 - 8 | 1605 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |