Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: smk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028738.7 | NP_001023909.2 | 8 - 9 | 2775 | 94 | |
NM_001028739.6 | NP_001023910.2 | 5 - 6 | 2163 | 94 | ||
NR_138495.1 | 7 - 8 | 2819 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |