Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: str-86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072919.2 | NP_505320.2 | 4 - 5 | 631 | 83 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |