Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: str-86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072919.2 | NP_505320.2 | 5 - 6 | 772 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |