Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srsx-34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380720.1 | NP_001367564.1 | 5 - 6 | 769 | 112 | |
NR_145827.1 | 13 - 14 | 1854 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |