Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oac-50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001322580.2 | NP_001309487.1 | 7 - 8 | 1086 | 120 | |
NR_136394.1 | 6 - 7 | 887 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |