Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ztf-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073125.6 | NP_505526.1 | 3 - 4 | 472 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |