Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: egl-47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028558.3 | NP_001023729.1 | 1 - 2 | 204 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |