Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: egl-47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028557.3 | NP_001023728.1 | 5 - 6 | 647 | 118 | |
NM_001028558.3 | NP_001023729.1 | 4 - 5 | 536 | 118 | ||
GenBank mRNA | AY532645.1 | - | 5 - 6 | 638 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |