Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srsx-26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383379.1 | 4 - 5 | 840 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |