Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcy-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307767.1 | NP_001294696.1 | 18 - 19 | 3177 | 69 | |
NM_073249.3 | NP_505650.3 | 18 - 19 | 3165 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |