Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ceeh-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269282.2 | NP_001256211.1 | 1 - 2 | 579 | 147 | |
GenBank mRNA | EU151492.1 | - | 1 - 2 | 341 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |