Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ugt-35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028523.1 | NP_001023694.1 | 4 - 5 | 946 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |