Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ugt-35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028523.1 | NP_001023694.1 | 6 - 7 | 1320 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |