Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srj-22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073338.2 | NP_505739.1 | 3 - 4 | 437 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |