Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srj-21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073339.3 | NP_505740.2 | 1 - 2 | 111 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |