Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: secs-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136415.3 | NP_001129887.1 | 4 - 5 | 382 | 77 | |
NM_073360.4 | NP_505761.1 | 3 - 4 | 256 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |