Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: secs-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136415.3 | NP_001129887.1 | 7 - 8 | 1306 | 102 | |
NM_073360.4 | NP_505761.1 | 6 - 7 | 1180 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |