Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F58B4.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383392.1 | 7 - 8 | 705 | 157 |
Gene Symbol: nas-31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028822.4 | NP_001023993.1 | 9 - 10 | 1159 | 157 | |
NM_001028823.6 | NP_001023994.1 | 9 - 10 | 1137 | 157 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |