Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hot-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073405.2 | NP_505806.1 | 1 - 2 | 129 | 121 | |
GenBank mRNA | AF324058.1 | - | 1 - 2 | 66 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Caenorhabditis elegans Probe spans exons |