Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lact-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269357.2 | NP_001256286.1 | 4 - 5 | 815 | 75 | |
NM_001269358.3 | NP_001256287.1 | 4 - 5 | 541 | 75 | ||
NM_001269359.3 | NP_001256288.1 | 2 - 3 | 193 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |