Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lact-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269357.2 | NP_001256286.1 | 6 - 7 | 1521 | 91 | |
NM_001269358.3 | NP_001256287.1 | 6 - 7 | 1247 | 91 | ||
NM_001269359.3 | NP_001256288.1 | 4 - 5 | 899 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |