Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srh-46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073470.3 | NP_505871.2 | 3 - 4 | 753 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |