Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F55C10.5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383404.1 | 2 - 3 | 221 | 109 |
Gene Symbol: cnb-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269388.2 | NP_001256317.1 | 3 - 4 | 271 | 109 | |
NM_001269389.2 | NP_001256318.1 | 3 - 4 | 271 | 109 | ||
GenBank mRNA | AY190131.1 | - | 2 - 3 | 222 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |