Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: deg-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073496.6 | NP_505897.1 | 4 - 5 | 1291 | 152 | |
GenBank mRNA | U19747.1 | - | 4 - 5 | 1313 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |