Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aat-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129542.4 | NP_001123014.1 | 5 - 6 | 1020 | 79 | |
NM_073504.5 | NP_505905.2 | 6 - 7 | 1115 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |