Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ccch-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073525.8 | NP_505926.2 | 7 - 8 | 1151 | 65 | |
NM_073526.5 | NP_505927.2 | 5 - 6 | 1196 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |