Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ape-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269424.2 | NP_001256353.1 | 2 - 3 | 221 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |