Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: daf-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073559.8 | NP_505960.3 | 3 - 4 | 489 | 144 | |
NR_132532.1 | 2 - 3 | 121 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |