Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rhr-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073560.6 | NP_505961.1 | 8 - 9 | 1067 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |