Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sel-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073568.6 | NP_505969.1 | 2 - 3 | 671 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |