Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srx-76 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073592.2 | NP_505993.2 | 3 - 4 | 469 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |