Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073606.5 | NP_506007.1 | 2 - 3 | 273 | 149 | |
NR_001507.1 | 2 - 3 | 201 | 149 | |||
GenBank mRNA | AY557405.1 | - | 2 - 3 | 230 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |