Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcy-33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269462.4 | NP_001256391.1 | 7 - 8 | 2123 | 75 | |
NM_001269463.3 | NP_001256392.1 | 7 - 8 | 2108 | 75 | ||
GenBank mRNA | AY275181.1 | - | 7 - 8 | 2102 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |