Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kup-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383418.1 | 12 - 13 | 1933 | 105 |
Gene Symbol: pkc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269466.3 | NP_001256395.1 | 5 - 6 | 612 | 105 | |
NM_001269467.3 | NP_001256396.1 | 4 - 5 | 573 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |