Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kup-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383418.1 | 17 - 18 | 2875 | 149 |
Gene Symbol: pkc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269465.2 | NP_001256394.1 | 6 - 7 | 1158 | 149 | |
NM_001269466.3 | NP_001256395.1 | 10 - 11 | 1554 | 149 | ||
NM_001269467.3 | NP_001256396.1 | 9 - 10 | 1515 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |