Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kup-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383418.1 | 4 - 5 | 394 | 159 | ||
GenBank mRNA | L12247.1 | - | 4 - 5 | 416 | 159 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |