Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kup-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383418.1 | 6 - 7 | 706 | 100 | ||
GenBank mRNA | L12247.1 | - | 6 - 7 | 728 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |