Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kup-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383418.1 | 7 - 8 | 937 | 85 | ||
GenBank mRNA | L12247.1 | - | 7 - 8 | 959 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |