Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clh-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073621.5 | NP_506022.1 | 8 - 9 | 2113 | 108 | |
GenBank mRNA | AF319616.1 | - | 8 - 9 | 2135 | 108 | |
AF328908.1 | - | 8 - 9 | 2144 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |