Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srj-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073650.1 | NP_506051.1 | 2 - 3 | 661 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |