Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lrx-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073671.6 | NP_506072.2 | 7 - 8 | 1192 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |