Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twk-24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380782.1 | NP_001367093.1 | 4 - 5 | 900 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |